Tisk…

tert-Butyl-4-aminobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: tert-Butyl 4-aminobenzoate

06975-25G 06975-5G
SIAL06975-25GEA 13810 CZK
SIAL06975-25G SIAL06975-5G
tert-Butyl-4-aminobenzoate, Sigma-Aldrich®
tert-Butyl-4-aminobenzoate
Vzorec: C₁₁H₁₅NO₂
MW: 193,2423 g/mol
MDL Number: MFCD00665790
CAS číslo: 18144-47-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR