Tisk…

2-Decyn-1-ol ≥97%

Dodavatel: Thermo Fisher Scientific

L05523.06 L05523.14
L05523.06EA 1970 CZK
L05523.06 L05523.14
2-Decyn-1-ol ≥97%
2-Decyn-1-ol

Purity: 97% CAS No.: 4117-14-0 Molecular Formula: CH3(CH2)6C≡CCH2OH Formula Weight: 154.25 Flash Point: 110°(230°F) Refractive Index: 1.4585 Boiling Point: 114-116°/6mm Beilstein Reference: 1852842 MDL No.: MFCD00041567

Vzorec: C₁₀H₁₈O
Bod varu: 114…116 °C (6 mmHg)
Hustota: 0,855
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041567
CAS číslo: 4117-14-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR