Tisk…
Contact plates
Dodavatel: CHERWELL
103070/E 103150/E
103070/EEA 0 CZK
103070/E 103150/E
Contact plates
Média mikrobiologická Misky mikrobiologické kontaktní
Contact plates 'Agar Contact Clean Room' are used either alone or with the 'Contact Weight' surface sampler for microbiological surface control and with SAS air samplers.
This type of contact plate is particularly useful for the validation of sterile chambers. Irradiated double or triple packing allows the insertion of the plates into the area to be monitored without any environmental contamination. By removing the most external protection before entering the white zone, it allows the material to enter the contaminated area.
It is possible to request custom formulations or packaging as needed.