Tisk…

Vinylphosphonic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Vinylphosphonicacid

M-Clarity™ quality level = MQ100

396311-25G 396311-5G
SIAL396311-25GEA 7850 CZK
SIAL396311-25G SIAL396311-5G
Vinylphosphonic acid, Sigma-Aldrich®
Vinylphosphonic acid
Vzorec: C₂H₅O₃P
MW: 108,0331 g/mol
MDL Number: MFCD00043866
CAS číslo: 1746-03-8
EINECS: 217-123-2
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR