Diethyl vinylphosphonate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
10368.04
10368.09
10368.04EA
2300
CZK
10368.04
10368.09
Diethyl vinylphosphonate ≥97%
Diethyl vinylphosphonate
Purity: 97%
CAS No.: 682-30-4
Molecular Formula: H2C=CHP(O)(OC2H5)2
Formula Weight: 164.14
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.068
Refractive Index: 1.4290
Boiling Point: 202°
UN No.: 3278
MDL No.: MFCD00009079
Vzorec:
C₆H₁₃O₃P MW: 164,14 g/mol Bod varu: 202 °C Hustota: 1,068 Bod vzplanutí: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009079 CAS číslo: 682-30-4 EINECS: 211-663-2 UN: 3278 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...