Tisk…

Diethyl vinylphosphonate 97%

Dodavatel: Thermo Scientific

152150100 152150250 152151000
ACRO152150100EA 5340 CZK
ACRO152150100 ACRO152150250 ACRO152151000
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate
Vzorec: C₆H₁₃O₃P
MW: 164,14 g/mol
Bod varu: 202 °C (760 mmHg)
Hustota: 1,06 g/cm³
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009079
CAS číslo: 682-30-4
EINECS: 211-663-2
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR