Tisk…

α-Naphtholphthalein

Dodavatel: Thermo Fisher Scientific
Varování

41221.03 41221.06 41221.14
41221.03EA 1970 CZK
41221.03 41221.06 41221.14
α-Naphtholphthalein
α-Naphtholphthalein

CAS No.: 596-01-0 Molecular Formula: C28H18O4 Formula Weight: 418.45 Physical Form: Powder Appearance: Pale brown Melting Point: 238-240° MDL No.: MFCD00036202


Indicator in 0.1% or 0.04% alcohol solution

Vzorec: C₂₈H₁₈O₄
Bod tání: 238…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00036202
CAS číslo: 596-01-0
EINECS: 209-875-5
Merck Index: 14,6389

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR