N-(4-Pentynyl)phthalimide, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(Pent-4-yn-1-yl)isoindoline-1,3-dione
M-Clarity™ quality level = MQ100
646938-25G
646938-5G
SIAL646938-25GEA
10280
CZK
SIAL646938-25G
SIAL646938-5G
N-(4-Pentynyl)phthalimide, Sigma-Aldrich®
N-(4-Pentynyl)phthalimide
Vzorec:
C₁₃H₁₁NO₂ MW: 213,24 g/mol |
MDL Number:
MFCD06798110 CAS číslo: 6097-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...