N-(4-Pentynyl)phthalimide ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(Pent-4-yn-1-yl)isoindoline-1,3-dione
H53432.06
H53432.14
H53432.06EA
2450
CZK
H53432.06
H53432.14
N-(4-Pentynyl)phthalimide ≥97%
N-(4-Pentynyl)phthalimide
Purity: 97%
CAS No.: 6097-07-0
Molecular Formula: C13H11NO2
Formula Weight: 213.24
Melting Point: 87-91°
MDL No.: MFCD06798110
Vzorec:
C₁₃H₁₁NO₂ MW: 213,24 g/mol Bod tání: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798110 CAS číslo: 6097-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...