Tisk…

N-(2-Butynyl)phthalimide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(But-2-yn-1-yl)isoindoline-1,3-dione

M-Clarity™ quality level = MQ100

663034-25G 663034-5G
SIAL663034-25GEA 7460 CZK
SIAL663034-25G SIAL663034-5G
N-(2-Butynyl)phthalimide, Sigma-Aldrich®
N-(2-Butynyl)phthalimide
Vzorec: C₁₂H₉NO₂
MW: 199.21 g/mol
MDL Number: MFCD00160830
CAS číslo: 113439-83-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR