Tisk…

Benzylmagnesium chloride ∼32% (w/w) 2 mol/l in THF, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

225916-100ML 225916-800ML
SIAL225916-100MLEA 3410 CZK
SIAL225916-100ML SIAL225916-800ML
Benzylmagnesium chloride ∼32% (w/w) 2 mol/l in THF, Sigma-Aldrich®
Benzylmagnesium chloride
Vzorec: C₇H₇ClMg
Storage Temperature: Ambient
MDL Number: MFCD00000469
CAS číslo: 6921-34-2
EINECS: 230-039-0
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR