Tisk…
Benzylmagnesium chloride ∼16% (w/w) 1 mol/l in diethyl ether, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
302759-800ML 302759-100ML
SIAL302759-800MLEA 8010 CZK
SIAL302759-800ML SIAL302759-100ML
Benzylmagnesium chloride ∼16% (w/w) 1 mol/l in diethyl ether, Sigma-Aldrich®
Benzylmagnesium chloride
Vzorec: C₇H₇ClMg
MW: 150.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000469
CAS číslo: 6921-34-2
EINECS: 230-039-0