Tisk…

Vinylphosphonic acid ≥90%, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Vinylphosphonicacid

M-Clarity™ quality level = MQ100

672068-1KG 672068-2.5KG 672068-250G
SIAL672068-1KGEA 0 CZK
SIAL672068-1KG SIAL672068-2.5KG SIAL672068-250G
Vinylphosphonic acid ≥90%, Sigma-Aldrich®
Vinylphosphonic acid
Vzorec: C₂H₅O₃P
MW: 108,0331 g/mol
MDL Number: MFCD00043866
CAS číslo: 1746-03-8
EINECS: 217-123-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR