Tisk…

Poly(2-ethyl-2-oxazoline), M.W. ~500,000, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

373974-100G 373974-500G
SIAL373974-100GEA 1760 CZK
SIAL373974-100G SIAL373974-500G
Poly(2-ethyl-2-oxazoline), M.W. ~500,000, Sigma-Aldrich®
Poly(2-ethyl-2-oxazoline)
Vzorec: (C₅H₉NO)x
MW: 200000 g/mol
CAS číslo: 25805-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR