Poly(2-ethyl-2-oxazoline), M.W. ~500,000
Dodavatel: Thermo Scientific
42398.22
42398.36
42398.A4
42398.22EA
1500
CZK
42398.22
42398.36
42398.A4
Poly(2-ethyl-2-oxazoline), M.W. ~500,000
Poly(2-ethyl-2-oxazoline)
CAS No.: 25805-17-8
Molecular Formula: [-N(COC2H5)CH2CH2-]n
Physical Form: Solid
MDL No.: MFCD00134377
Vzorec:
(C₅H₉NO)x MW: 200000 g/mol Storage Temperature: Ambient |
CAS číslo:
25805-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...