Tisk…
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 2,2-Dimethyl-1,3-dioxane-4,6-dione
M-Clarity™ quality level = MQ200
210145-100G 210145-10G 210145-25G
SIAL210145-100GEA 4040 CZK
SIAL210145-100G SIAL210145-10G SIAL210145-25G
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid), Sigma-Aldrich®
2,2-Dimethyl-1,3-dioxane-4,6-dione (isopropylidene malonate, meldrum's acid)
Vzorec: C₆H₈O₄
MW: 144.13 g/mol
MDL Number: MFCD00006638
CAS číslo: 2033-24-1
EINECS: 217-992-8