Tisk…

Methyl-6-bromo-2-naphthoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 6-bromo-2-naphthoate

M-Clarity™ quality level = MQ100

539406-25G 539406-5G
SIAL539406-25GEA 8530 CZK
SIAL539406-25G SIAL539406-5G
Methyl-6-bromo-2-naphthoate, Sigma-Aldrich®
Methyl-6-bromo-2-naphthoate
Vzorec: C₁₂H₉BrO₂
MW: 265,11 g/mol
MDL Number: MFCD00100408
CAS číslo: 33626-98-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR