Benzyl isocyanide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
133299-1G
133299-5G
SIAL133299-1GEA
3150
CZK
SIAL133299-1G
SIAL133299-5G
Benzyl isocyanide, Sigma-Aldrich®
Benzyl isocyanide
Vzorec:
C₈H₇N MW: 117,15 g/mol |
MDL Number:
MFCD00000004 CAS číslo: 10340-91-7 EINECS: 233-738-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...