Tisk…

4-(Pentafluorothio)phenol ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H33413.03 H33413.06
H33413.03EA 2970 CZK
H33413.03 H33413.06
4-(Pentafluorothio)phenol ≥97%
4-(Pentafluorothio)phenol

Purity: 97% CAS No.: 774-94-7 Molecular Formula: C6H5F5OS Formula Weight: 220.16 MDL No.: MFCD03788516

Vzorec: C₆H₅F₅OS
Storage Temperature: Ambient
MDL Number: MFCD03788516
CAS číslo: 774-94-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR