Tisk…

3-Fluoro-5-(pentafluorothio)aniline ≥97%

Dodavatel: Thermo Scientific
Varování

H33680.MD H33680.03
H33680.MDEA 4880 CZK
H33680.MD H33680.03
3-Fluoro-5-(pentafluorothio)aniline ≥97%
3-Fluoro-5-(pentafluorothio)aniline

Purity: 97% Molecular Formula: C6H5F6NS Formula Weight: 237.16 UN No.: 2811 MDL No.: MFCD16652430

Vzorec: C₆H₅F₆NS
MW: 237,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652430
CAS číslo: 1240256-99-8
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR