Tisk…

2-Fluoro-4-(pentafluorothio)aniline ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H34099.MD H34099.03
H34099.MDEA 6800 CZK
H34099.MD H34099.03
2-Fluoro-4-(pentafluorothio)aniline ≥97%
2-Fluoro-4-(pentafluorothio)aniline

Purity: 97% CAS No.: 1240257-25-3 Molecular Formula: C6H5F6NS Formula Weight: 237.16 UN No.: 2810 MDL No.: MFCD16652511

Vzorec: C₆H₅F₆NS
Storage Temperature: Ambient
MDL Number: MFCD16652511
CAS číslo: 1240257-25-3
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR