Tisk…

4-[(Trifluoromethyl)thio]aniline ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-((Trifluoromethyl)thio)aniline

L16067.03 L16067.06 L16067.14
L16067.03EA 848 CZK
L16067.03 L16067.06 L16067.14
4-[(Trifluoromethyl)thio]aniline ≥98%
4-[(Trifluoromethyl)thio]aniline

Purity: 98% CAS No.: 372-16-7 Molecular Formula: C7H6F3NS Formula Weight: 193.19 Flash Point: 106°(222°F) Refractive Index: 1.5280 Odour: Unpleasant Boiling Point: 103°/11mm Beilstein Reference: 2208859 UN No.: 2810 MDL No.: MFCD00040926

Vzorec: C₇H₆F₃NS
MW: 193.19 g/mol
Bod varu: 103 °C (11 mmHg)
Hustota: 1.344
Bod vzplanutí: 106 °C (222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040926
CAS číslo: 372-16-7
EINECS: 206-744-4
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR