4-Acetamidobenzenesulphonyl azide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
404764-25G
404764-5G
SIAL404764-25GEA
4250
CZK
SIAL404764-25G
SIAL404764-5G
4-Acetamidobenzenesulphonyl azide, Sigma-Aldrich®
4-Acetamidobenzenesulphonyl azide
Vzorec:
C₈H₈N₄O₃S MW: 240,24 g/mol |
MDL Number:
MFCD00029626 CAS číslo: 2158-14-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...