Tisk…

4-(4-Chlorophenoxy)aniline, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Amino-4'-chlorodiphenyl ether

M-Clarity™ quality level = MQ100

695386-25G 695386-5G
SIAL695386-25GEA 6990 CZK
SIAL695386-25G SIAL695386-5G
4-(4-Chlorophenoxy)aniline, Sigma-Aldrich®
4-(4-Chlorophenoxy)aniline
Vzorec: C₁₂H₁₀ClNO
MW: 219,67 g/mol
MDL Number: MFCD00043925
CAS číslo: 101-79-1
EINECS: 202-976-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR