Methyl-2-amino-5-chloro-3-iodobenzoate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
652628-25G
652628-5G
SIAL652628-25GEA
0
CZK
SIAL652628-25G
SIAL652628-5G
Methyl-2-amino-5-chloro-3-iodobenzoate, Sigma-Aldrich®
Methyl-2-amino-5-chloro-3-iodobenzoate
Vzorec:
C₈H₇ClINO₂ MW: 311,51 g/mol |
MDL Number:
MFCD00673017 CAS číslo: 289039-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...