(4-Acetamidophenyl)boronic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Acetamidophenylboronic acid, 4-Acetamidobenzeneboronic acid, 4-Acetylaminophenylboronic acid
H52987.03
H52987.06
H52987.03EA
3780
CZK
H52987.03
H52987.06
(4-Acetamidophenyl)boronic acid 96%
(4-Acetamidophenyl)boronic acid
Purity: 96%
CAS No.: 101251-09-6
Molecular Formula: C8H10BNO3
Formula Weight: 178.98
Melting Point: 215-220°
MDL No.: MFCD02179451
Vzorec:
C₈H₁₀BNO₃ Bod tání: 215…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179451 CAS číslo: 101251-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...