4-Mercaptophenylacetic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
653152-1G
653152-5G
SIAL653152-1GEA
3070
CZK
SIAL653152-1G
SIAL653152-5G
4-Mercaptophenylacetic acid, Sigma-Aldrich®
4-Mercaptophenylacetic acid
Vzorec:
C₈H₈O₂S |
MDL Number:
MFCD00797617 CAS číslo: 39161-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...