Tisk…

4-Mercaptophenylacetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H27658.06 H27658.03
H27658.06EA 8140 CZK
H27658.06 H27658.03
4-Mercaptophenylacetic acid ≥97%
4-Mercaptophenylacetic acid

Purity: 97% CAS No.: 39161-84-7 Molecular Formula: C8H8O2S Formula Weight: 168.21 Storage Temperature: 2°C to 8°C Melting Point: 105-110° MDL No.: MFCD00797617

Vzorec: C₈H₈O₂S
Bod tání: 105…110 °C
MDL Number: MFCD00797617
CAS číslo: 39161-84-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR