Tisk…

Potassium tetraoxalate dihydrate 99%, extra pure

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate

352111000 352110010 352115000
ACRO352111000EA 1010 CZK
ACRO352111000 ACRO352110010 ACRO352115000
Potassium tetraoxalate dihydrate 99%, extra pure
Potassium tetraoxalate dihydrate
Vzorec: C₄H₃KO₈·2H₂O
MW: 254.19 g/mol (218.16 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150443
CAS číslo: 6100-20-5
EINECS: 204-874-6

Objednat

Výsledky specifikačního testu

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying 12 % to 16 % (1 g, 105°C)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR