L(+)-Glutamin 0.2 mol/l in aqueous solution, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
M-Clarity™ quality level = MQ300
G7513-100ML
G7513-20ML
SIALG7513-100MLEA
1130
CZK
SIALG7513-100ML
SIALG7513-20ML
L(+)-Glutamin 0.2 mol/l in aqueous solution, Sigma-Aldrich®
L(+)-Glutamin
Vzorec:
C₅H₁₀N₂O₃ MW: 146.15 Bod varu: 355 Storage Temperature: Ambient |
MDL Number:
MFCD00008044 CAS číslo: 56-85-9 EINECS: 200-292-1 Merck Index: 13,04484 |