Ammonium 8-anilinonaphthalene-1-sulphonate hydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
216909-250G
216909-5G
SIAL216909-250GEA
25210
CZK
SIAL216909-250G
SIAL216909-5G
Ammonium 8-anilinonaphthalene-1-sulphonate hydrate, Sigma-Aldrich®
Ammonium 8-anilinonaphthalene-1-sulphonate hydrate
Vzorec:
C₁₆H₁₆N₂O₃S·H₂O |
CAS číslo:
206659-00-9 EINECS: 249-265-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...