Benzo[b]furan-2-boronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Benzofuran-2-boronic acid, 2-Benzofurylboronic acid
B23676.03
B23676.06
B23676.14
B23676.03EA
1530
CZK
B23676.03
B23676.06
B23676.14
Benzo[b]furan-2-boronic acid ≥98%
Benzo[b]furan-2-boronic acid
Purity: 98%
CAS No.: 98437-24-2
Molecular Formula: C8H7BO3
Formula Weight: 161.95
Storage Temperature: 2°C to 8°C
Vzorec:
C₈H₇BO₃ MW: 161,95 g/mol |
MDL Number:
MFCD00236019 CAS číslo: 98437-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...