Tisk…

5-Methylfuran-2-boronic acid pinacol ester ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan, (5-Methylfuran-2-yl)boronic acid Pinacol ester

H31282.03 H31282.06
H31282.03EA 783 CZK
H31282.03 H31282.06
5-Methylfuran-2-boronic acid pinacol ester ≥97%
5-Methylfuran-2-boronic acid pinacol ester

Purity: 97% CAS No.: 338998-93-9 Molecular Formula: C11H17BO3 Formula Weight: 208.06 MDL No.: MFCD03094689

Vzorec: C₁₁H₁₇BO₃
MW: 208.06 g/mol
Hustota: 1.023
Storage Temperature: Ambient
MDL Number: MFCD03094689
CAS číslo: 338998-93-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR