Tisk…

Furan-3-boronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(3-Furanyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan

M-Clarity™ quality level = MQ100

575054-1G 575054-5G
SIAL575054-1GEA 3350 CZK
SIAL575054-1G SIAL575054-5G
Furan-3-boronic acid pinacol ester, Sigma-Aldrich®
Furan-3-boronic acid pinacol ester
Vzorec: C₁₀H₁₅BO₃
MW: 194,035 g/mol
MDL Number: MFCD03453669
CAS číslo: 248924-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR