Tisk…

3,4-(Methylenedioxy)phenylacetonitrile ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L09518.09 L09518.18
L09518.09EA 2270 CZK
L09518.09 L09518.18
3,4-(Methylenedioxy)phenylacetonitrile ≥98%
3,4-(Methylenedioxy)phenylacetonitrile

Purity: 98+% CAS No.: 4439-02-5 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Flash Point: 110°(230°F) Melting Point: 41-45° Boiling Point: 135-140°/5mm Beilstein Reference: 7739 UN No.: 3439 MDL No.: MFCD00005835

Vzorec: C₉H₇NO₂
Bod varu: 135…140 °C (5 mmHg)
Bod tání: 41…45 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005835
CAS číslo: 4439-02-5
EINECS: 224-655-9
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR