Tisk…

N-(Phenylseleno)phthalimide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

254614-1G 254614-5G
SIAL254614-1GEA 1960 CZK
SIAL254614-1G SIAL254614-5G
N-(Phenylseleno)phthalimide, Sigma-Aldrich®
N-(Phenylseleno)phthalimide
Vzorec: C₁₄H₉NO₂Se
MDL Number: MFCD00010406
CAS číslo: 71098-88-9
UN: 3283
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR