Tisk…

Tetramethyl methylenebisphosphonate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13441.06 A13441.14 A13441.22
A13441.06EA 3810 CZK
A13441.06 A13441.14 A13441.22
Tetramethyl methylenebisphosphonate ≥98%
Tetramethyl methylenebisphosphonate

Purity: 98+% CAS No.: 16001-93-7 Molecular Formula: C5H14O6P2 Formula Weight: 232.11 Refractive Index: 1.4550 Boiling Point: 152-154°/2.5mm MDL No.: MFCD00014884

Vzorec: C₅H₁₄O₆P₂
Bod varu: 152…154 °C (2,5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00014884
CAS číslo: 16001-93-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR