Tisk…

Tetraisopropyl methylenebisphosphonate ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Tetraisopropyl methylenebis(phosphonate), Tetraisopropyl methylenediphosphonate

A13484.09 A13484.18
A13484.09EA 2190 CZK
A13484.09 A13484.18
Tetraisopropyl methylenebisphosphonate ≥98%
Tetraisopropyl methylenebisphosphonate

Purity: 98% CAS No.: 1660-95-3 Molecular Formula: C13H30O6P2 Formula Weight: 344.33 Flash Point: 160°(320°F) Refractive Index: 1.4340 Boiling Point: 122-124°/1.5mm Beilstein Reference: 1080180 MDL No.: MFCD00015021

Vzorec: C₁₃H₃₀O₆P₂
MW: 344,33 g/mol
Bod varu: 122…124 °C (1,5 mmHg)
Hustota: 1,080
Bod vzplanutí: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015021
CAS číslo: 1660-95-3
EINECS: 216-765-0
UN: 3278
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR