Tisk…

Tetraethyl ethylenebisphosphonate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Tetraethyl ethylenediphosphonate, Tetraethyl ethane-1,2-diylbis(phosphonate)

A14206.03 A14206.06 A14206.14
A14206.03EA 851 CZK
A14206.03 A14206.06 A14206.14
Tetraethyl ethylenebisphosphonate ≥98%
Tetraethyl ethylenebisphosphonate

Purity: 98% CAS No.: 995-32-4 Molecular Formula: C10H24O6P2 Formula Weight: 302.25 Flash Point: 110°(230°F) Refractive Index: 1.4410 Boiling Point: 153-154°/0.9mm Beilstein Reference: 1800096 MDL No.: MFCD00015133

Vzorec: C₁₀H₂₄O₆P₂
MW: 302,25 g/mol
Bod varu: 153…154 °C (0,9 mmHg)
Hustota: 1,146
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015133
CAS číslo: 995-32-4
EINECS: 213-625-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR