Diethyl (2-oxopropyl)phosphonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Diethyl acetylmethylphosphonate
370479-25G
370479-5G
SIAL370479-25GEA
12760
CZK
SIAL370479-25G
SIAL370479-5G
Diethyl (2-oxopropyl)phosphonate, Sigma-Aldrich®
Diethyl (2-oxopropyl)phosphonate
Vzorec:
C₇H₁₅O₄P MW: 194.17 g/mol |
MDL Number:
MFCD00044728 CAS číslo: 1067-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...