Tisk…

Ethyl-4-piperidinecarboxylate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

E33505-100G E33505-25G
SIALE33505-100GEA 5820 CZK
SIALE33505-100G SIALE33505-25G
Ethyl-4-piperidinecarboxylate, Sigma-Aldrich®
Ethyl-4-piperidinecarboxylate
Vzorec: C₈H₁₅NO₂
MDL Number: MFCD00006003
CAS číslo: 1126-09-6
EINECS: 214-416-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR