Diethyl (4-methylbenzyl)phosphonate ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Diethyl 4-methylbenzylphosphonate
A15190.09
A15190.18
A15190.09EA
1110
CZK
A15190.09
A15190.18
Diethyl (4-methylbenzyl)phosphonate ≥98%
Diethyl (4-methylbenzyl)phosphonate
Purity: 98+%
CAS No.: 3762-25-2
Molecular Formula: C12H19O3P
Formula Weight: 242.26
Flash Point:
Vzorec:
C₁₂H₁₉O₃P MW: 242,26 g/mol Bod varu: 146…148 °C (0,6 mmHg) Hustota: 1,070 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041385 CAS číslo: 3762-25-2 EINECS: 223-180-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...