Tisk…

Triethyl-2-phosphonoppentanoate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Ethyl 2-(diethoxyphosphoryl)pentanoate

30413.04 30413.09
30413.04EA 4260 CZK
30413.04 30413.09
Triethyl-2-phosphonoppentanoate ≥98%
Triethyl-2-phosphonoppentanoate

Purity: 98% CAS No.: 35051-49-1 Molecular Formula: C11H23O5P Formula Weight: 266.27 Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00015163

Vzorec: C₁₁H₂₃O₅P
MW: 266,27 g/mol
Bod varu: 98 °C (2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00015163
CAS číslo: 35051-49-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR