Tisk…

Triethyl-2-phosphonopropionate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate

M-Clarity™ quality level = MQ100

174653-10G 174653-50G
SIAL174653-10GEA 2180 CZK
SIAL174653-10G SIAL174653-50G
Triethyl-2-phosphonopropionate, Sigma-Aldrich®
Triethyl-2-phosphonopropionate
Vzorec: C₉H₁₉O₅P
MW: 238.22 g/mol
MDL Number: MFCD00009159
CAS číslo: 3699-66-9
EINECS: 223-033-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR