Tisk…

(3,4,5-Trimethoxyphenyl)acetonitrile ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B24247.06 B24247.14
B24247.06EA 1500 CZK
B24247.06 B24247.14
(3,4,5-Trimethoxyphenyl)acetonitrile ≥97%
(3,4,5-Trimethoxyphenyl)acetonitrile

Purity: 97% CAS No.: 13338-63-1 Molecular Formula: C11H13NO3 Formula Weight: 207.23 Melting Point: 75-79° Beilstein Reference: 2214548 UN No.: 3439 MDL No.: MFCD00001912

Vzorec: C₁₁H₁₃NO₃
Bod tání: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001912
CAS číslo: 13338-63-1
EINECS: 236-388-5
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR