Tisk…

Disodium maleate, Sigma-Aldrich®

Dodavatel: Merck

63260-250G 63260-50G
SIAL63260-250GEA 11930 CZK
SIAL63260-250G SIAL63260-50G
Disodium maleate, Sigma-Aldrich®
Disodium maleate
Vzorec: C₄H₂Na₂O₄
MDL Number: MFCD00013059
CAS číslo: 371-47-1
EINECS: 206-738-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR