Tisk…

4-Chloro-7-fluoro-2-methylquinoline 95%

Dodavatel: Thermo Fisher Scientific
Varování

H50554.03 H50554.06
H50554.03EA 2640 CZK
H50554.03 H50554.06
4-Chloro-7-fluoro-2-methylquinoline 95%
4-Chloro-7-fluoro-2-methylquinoline

Purity: 95% CAS No.: 18529-04-9 Molecular Formula: C10H7ClFN Formula Weight: 195.63 MDL No.: MFCD00272333

Vzorec: C₁₀H₇ClFN
MW: 195.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272333
CAS číslo: 18529-04-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR