(S)-(+)-Nipecotic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
L-Nipecotic acid, (S)-(+)-3-Piperidinecarboxylic acid, (S)-Piperidine-3-carboxylic acid
M-Clarity™ quality level = MQ100
656364-10G
656364-1G
SIAL656364-10GEA
7740
CZK
SIAL656364-10G
SIAL656364-1G
(S)-(+)-Nipecotic acid, Sigma-Aldrich®
(S)-(+)-Nipecotic acid
Vzorec:
C₆H₁₁NO₂ MW: 129,16 g/mol |
MDL Number:
MFCD01630807 CAS číslo: 59045-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...