(S)-(+)-Nipecotic acid ≥96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
L-Nipecotic acid, (S)-(+)-3-Piperidinecarboxylic acid, (S)-Piperidine-3-carboxylic acid
H57543.06
H57543.14
H57543.06EA
3570
CZK
H57543.06
H57543.14
(S)-(+)-Nipecotic acid ≥96%
(S)-(+)-Nipecotic acid
Purity: 96+%
CAS No.: 59045-82-8
Molecular Formula: C6H11NO2
Formula Weight: 129.16
Melting Point: 253-257°
Optical Rotation: +5° (c=1 in water)
Vzorec:
C₆H₁₁NO₂ MW: 129,16 g/mol Bod tání: 253…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD01630807 CAS číslo: 59045-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...