Tisk…

(R)-(-)-Nipecotic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid

M-Clarity™ quality level = MQ100

656356-10G 656356-1G
SIAL656356-10GEA 7700 CZK
SIAL656356-10G SIAL656356-1G
(R)-(-)-Nipecotic acid, Sigma-Aldrich®
(R)-(-)-Nipecotic acid
Vzorec: C₆H₁₁NO₂
MW: 129,16 g/mol
MDL Number: MFCD01630787
CAS číslo: 25137-00-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR